About this item
LightShift® Chemiluminescent EMSA Kit is robust and sensitive system for performing electrophoretic mobility shift assays (EMSAs) to identify and characterize protein:DNA binding interactions.
- Excellent for detecting low-abundance proteins in nuclear extracts
- Sensitivity that surpasses radioactive and digoxigenin methods
- Compatible with previously established binding conditions for popular DNA-protein interactions
- Includes EBNA Optimization and Control Kit to help new users develop a working assay and understand the methods used to confirm binding interaction specificity
The EMSA technique is based on the observation that protein:DNA complexes migrate more slowly than free DNA molecules when subjected to non-denaturing polyacrylamide or agarose gel electrophoresis. Because the rate of DNA migration is shifted or retarded upon protein binding, the assay is also referred to as a gel shift or gel retardation assay. DNA targets can be synthesized with 5' or 3' biotin labels or they can be labeled after synthesis using the Thermo Scientific Biotin 3' End DNA Labeling Kit (Product No. 89818). Nuclear, cytosolic or whole cell protein extracts can be obtained by a variety of methods, including the Thermo Scientific NE-PER Nuclear and Cytoplasmic Extraction Reagent Kit.