BD BBL™ Trypticase Soy Agar (TSA) Plates, BD Diagnostics
Catalog # 90006-556
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Prepared Plates
- Description:BD Trypticase™ Soy Agar with Lecithin and Polysorbate 80, Double-Wrapped
- Cat. No.:90006-556
- Plate size:100×15 mm
- Supplier no.:221943
Specifications
About this item
Trypticase Soy Agar (TSA) plates, pre-filled and lot-to-lot tested.
- Pre-filled plates
- Lot controlled
- Reproducible results
The growth characteristics of these pre-filled plates are carefully controlled, and samples of each lot are tested. Each lot passes a rigorous battery of quality tests including biological growth promotion and physical characteristics analysis, which ensures accurate, reproducible results. BD RODAC™ agar plates with a convex bottom and wide edge are specifically designed for surface monitoring while keeping the agar bed in place during transport and use.