Order Entry
United States
ContactUsLinkComponent
2-Butenyl-4-methyl-threonine (BMT) 0.3M in acetonitrile with 0.5% N-Methylimidazole, BioSyn™ activator reagent, for DNA/RNA synthesis, Burdick & Jackson™
Health
Irritant
Flammable
2-Butenyl-4-methyl-threonine (BMT) 0.3M in acetonitrile with 0.5% N-Methylimidazole, BioSyn™ activator reagent, for DNA/RNA synthesis, Burdick & Jackson™
Catalog # BJNP729-20L
CAS Number:  
Health
Irritant
Flammable
2-Butenyl-4-methyl-threonine (BMT) 0.3M in acetonitrile with 0.5% N-Methylimidazole, BioSyn™ activator reagent, for DNA/RNA synthesis, Burdick & Jackson™
Catalog # BJNP729-20L
Supplier Number:  NP729-20L
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Returnable Stainless Steel Drum
  • Size:
    20 L
  • Cat. No.:
    BJNP729-20L

Specifications

About this item

 :
UN:1648
 :GHS08
 :GHS07
 :GHS02
Boiling Pt:82 °C
Density:0.8083 g/cm3 at 20 °C
Flash Pt:52 °F (11 °C)
Storage Temperature:Ambient
ADR:3,II
 :187.24 g/mol
CAS Number:81135-57-1
Formula:C₉H₁₇NO₃