ContactUsLinkComponent
Fundamentals of Physics Infinite Photogate Adapter
Fundamentals of Physics Infinite Photogate Adapter
Catalog # 470352-192
Supplier:  Avantor
undefined
Fundamentals of Physics Infinite Photogate Adapter
Catalog # 470352-192
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Fundamentals of physics infinite photogate adapter
  • Cat. No.:
    470352-192
Specifications
About this item
Extend your photogate to accommodate any width.
  • Clips securely to the Fundamentals of Physics Photogate system
  • Two mirrored adapters allow both photogates to be set up with extra long gate lengths
  • Perfect for a wide variety of physics experiments, such as free fall, uniform circular motion, and attwood machines
Want to analzye the motion of an object that is too large to fit through the arms of a photogate? Whether it's a tennis ball in free fall or a student-constructed sail cart, this simple device allows you to create a photogate with an 'infinite' path length. The mirrored clips attach easily and securely to the infrared 'source' side of the photogate. A red laser pointer can then be aimed at the mirror so that the beam reflects into the detector side of the photogate. If an object blocks the laser's beam, the photogate will register an 'event' as usual.
Accessories information: At least one red laser pointer is required but available separately.
Packaging: Mirrors are shipped with a protective film to prevent scratching. Remove film before use.