Tissue-Tek® Processing/Embedding Cassette Systems, Sakura
About this item
Cassette systems (cassette, cover, and base) are designed for processing, embedding, sectioning, and storage
Reusable base molds enable perfect specimen positioning. Extended fins prevent tipping and provide gripping surface. Stainless steel construction ensures uniform heating and cooling.
Note: Clear cassettes are polypropylene and are not recommended for use in processors using heat or vacuum.
Plastic cassettes feature an angled, textured writing surface. Available in clear and in 10 colors.
Ordering information: Reusable stainless steel cover sold separately.
10 Options Available Below
Product Details & Documents
Cassette systems (cassette, cover, and base) are designed for processing, embedding, sectioning, and storage
Reusable base molds enable perfect specimen positioning. Extended fins prevent tipping and provide gripping surface. Stainless steel construction ensures uniform heating and cooling.
Note: Clear cassettes are polypropylene and are not recommended for use in processors using heat or vacuum.
Plastic cassettes feature an angled, textured writing surface. Available in clear and in 10 colors.
Ordering information: Reusable stainless steel cover sold separately.
Product Family Options
Product Information
- ColorAvailabilityPrice
- Supplier Number: 4126
- Supplier Number: 4179
- Supplier Number: 4182
- Supplier Number: 4127
- Supplier Number: 4191
- Supplier Number: 4125
- Supplier Number: 4129
- Supplier Number: 4183
- Supplier Number: 4184
- Supplier Number: 4187
Accessories
Product Information
- Catalog #LengthWidthAvailabilityPrice
- Base Mold
Product Information
- Catalog #LengthWidthAvailabilityPrice
- Base Mold
Product Information
- Catalog #LengthWidthAvailabilityPrice
- Base Mold
Product Information
- Catalog #LengthWidthAvailabilityPrice
- Base Mold
Product Information
- Catalog #AvailabilityPrice
- Stainless Steel Cover
Product Information
- Catalog #LengthWidthAvailabilityPrice
- Base Mold