Order Entry
United States
ContactUsLinkComponent
Chiron Per- and Polyfluoroalkyl Substances (PFAS) Standards in Methanol
Chiron Per- and Polyfluoroalkyl Substances (PFAS) Standards in Methanol
Supplier:  SPEX CERTIPREP LLC
Certificates
undefined
Chiron Per- and Polyfluoroalkyl Substances (PFAS) Standards in Methanol
Supplier:  SPEX CERTIPREP LLC

Specifications

  • CAS No.:
    41997-13-1
  • Storage Conditions:
    Protect from air and light
  • Shipping Conditions:
    Ambient
  • Synonym(s):
    FHSA
  • Shelf Life:
    12 Months
  • Storage Temperature:
    −20 ±5 °C
  • Description:
    n-Perfluorohexanesulfonamide
  • Physical Form:
    Liquid

Specifications

Frequently Bought Together