NSI Lab Solutions Water Chemistry CRMS, Alkalinity Hardness
Catalog # SXQCI-041
Supplier: SPEX CERTIPREP LLC
New Product
CAS Number:
Specifications
- Matrix:Water
- Pack type:Plastic bottle
- CAS No.:7757-82-6, 1303-96-4, 7681-49-4, 7757-79-1
- Description:Sample for determination of chloride, conductivity, fluoride, nitrate as N, potassium, sulfate, nitrate/nitrite-N, alkalinity, total dissolved solids
- Potassium:Yes
- Fluoride:Yes
- Nitrate plus Nitrite:Yes
- Storage Temperature:15...30 °C
- Alkalinity:Yes
- Chloride:Yes
- Conductivity:Yes
- Nitrate:Yes
- Sulphate:Yes
- TDS:Yes
- Cat. No.:SXQCI-041
- Range:Potassium 10 - 40 mg/L, nitrate-N 3 - 10 mg/L, nitrate/nitrite-N 3 - 10 mg/L, chloride 20 - 160 mg/L, sulfate 25 - 250 mg/L, fluoride 1 - 8 mg/L, TDS at 180 °C 100 - 1000 mg/L, conductivity 130 - 1300 µm hos/cm, alkalinity 25 - 200 mg/L
- Volume:500 ml
Specifications
About this item
We have standards to support your water chemistry testing.
- Validate the accuracy of your water chemistry testing methods
- Manufactured and certified under ISO 17034 accreditation
- Meets your ISO 17025 and ISO 9001 requirements
Accuracy, homogeneity, and performance of each lot is certified in our ISO 17025 accredited laboratory by the same methods you use.