ProMTag Intact Protein Universal Protein Cleanup Kit, Impact Proteomics
Catalog # MSPP-WP-2000
Supplier: IMPACT PROTEOMICS, LLC MS
CAS Number:
Specifications
- Description:ProMTag Intact Protein Universal Protein Cleanup Kit
- Size:1 kit
- Cat. No.:MSPP-WP-2000
- Details:Protein amount per sample 1 – 40 µg
- Sufficient for:8 samples
Specifications
About this item
The ProMTag Intact Protein Universal Protein Cleanup Kit produces high-quality intact protein samples for mass spectrometry analysis or other downstream applications.
- Compatible with most starting samples including cultured cells, tissues, biological fluids, and pre-made lysates so long as they do not contain buffers with primary amines (such as TRIS).
- All reagents and plasticware for 8 sample preparations are included.
- Requires only standard laboratory equipment.
The ProMTag Intact Protein Universal Protein Cleanup Kit utilizes the reversible chemistry of ProMTag to produce high-quality protein samples for mass spectrometry analysis or other downstream applications in a simple, fast workflow with minimal hands-on time. Works with any starting sample for rapid, high yield extraction and cleanup of protein samples. Each kit contains reagents for 8 sample preparations.