Order Entry
United States
ContactUsLinkComponent
ProMTag Immunoprecipitation to Mass Spectrometry (IP-to-MS) Kit, Impact Proteomics
ProMTag Immunoprecipitation to Mass Spectrometry (IP-to-MS) Kit, Impact Proteomics
Catalog # MSPP-IP-1000
CAS Number:  
undefined
ProMTag Immunoprecipitation to Mass Spectrometry (IP-to-MS) Kit, Impact Proteomics
Catalog # MSPP-IP-1000
Supplier Number:  IP-1000
CAS Number:  

Specifications

  • Description:
    ProMTag Immunoprecipitation to Mass Spectrometry (IP-to-MS) Kit
  • Size:
    1 kit
  • Cat. No.:
    MSPP-IP-1000
  • Details:
    Protein amount per sample: 100 µg protein, 2 µL antibody or 10 µL plasma/serum
  • Sufficient for:
    8 samples

Specifications

About this item

The ProMTag Immunoprecipitation to Mass Spectrometry (IP-to-MS) Kit produces high-quality antigen samples ready for mass spectrometry analysis.

  • Compatible with a variety of antibody sources including purified antibodies, serum, plasma, bronchoalveolar lavage fluid, and cerebrospinal fluid.
  • Reagents and plasticware for 8 sample preparations are included. User provides protease inhibitors and antibody and antigen sources.
  • Includes Impact Proteomics' MT-Trypsin. The user does not need to provide an enzyme.
  • Requires only standard laboratory equipment.

The ProMTag Immunoprecipitation to Mass Spectrometry (IP-to-MS) Kit produces high-quality antigen samples for mass spectrometry analysis in a fast, simple workflow. Purify target antigens without worrying about contaminating antibodies, detergents, or salts. Each kit contains reagents for 8 immunoprecipitations.