Order Entry
United States
ContactUsLinkComponent
 
ProMTag Multiomics DNA, RNA, Protein Sample Preparation Kit, Impact Proteomics
Catalog # MSPP-MULTI1000
CAS Number:  
undefined
ProMTag Multiomics DNA, RNA, Protein Sample Preparation Kit, Impact Proteomics
Catalog # MSPP-MULTI1000
Supplier Number:  MULTI-1000
CAS Number:  

Specifications

  • Description:
    ProMTag Multiomics DNA, RNA, Protein Sample Preparation Kit
  • Size:
    1 kit
  • Environmentally Preferable:
  • Cat. No.:
    MSPP-MULTI1000
  • Details:
    Protein amount per sample: 1-150 µg protein, or 5-10 mg tissue
  • Sufficient for:
    8 samples

Specifications

About this item

The ProMTag Multiomics DNA, RNA, Protein Sample Preparation kit produces high-quality DNA, RNA, and peptide samples from the same starting sample.

  • Compatible with most starting samples including cultured cells, tissues, and pre-made lysates so long as they do not contain buffers with primary amines (such as TRIS).
  • Reagents and plasticware for 8 sample preparations are included. User provides RNase and/or DNase depending on downstream applications and RNase inhibitors if necessary.
  • Includes Impact Proteomics' MT-Trypsin. The user does not need to provide an enzyme.
  • Requires only standard laboratory equipment.

The ProMTag Multiomics DNA, RNA, Protein Sample Preparation kit utilizes the reversible chemistry of ProMTag to produce high-quality DNA, RNA, and peptide samples from the same starting sample. DNA and RNA are compatible with downstream approaches such as whole genome sequencing and RNA-Seq. Peptides are ready for mass spectrometry analysis. Included reagents and an easy-to-perform workflow take you from cell or tissue lysis all the way to DNA, RNA, and peptides. Each kit contains reagents for 8 sample preparations.