Toluidine blue Penetrant Solution ASTM F1929
Catalog # RCR8656000-1A
Supplier: Ricca Chemical
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:1 L
- Cat. No.:RCR8656000-1A
Specifications
Specification Test Results
- Appearance:Blue-purple liquid
- UV/Visible Spectrum:To Pass Test
Specification Test Results
About this item
:
:747.96
Storage Temperature:Ambient
CAS Number:6586-04-5
Formula:C₁₅H₁₆ClN₃S·0.5ZnCl₂
MDL Number:MFCD03548293