
Sodium sulfide solution Reag. Ph. Eur. 1083902 (R1)
Catalog # RCR7565000-1C
Supplier: Ricca Chemical
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:1 L
- Cat. No.:RCR7565000-1C
Specifications
Specification Test Results
- Appearance:Colorless to yellow liquid
Specification Test Results
About this item
:
Storage Temperature:Ambient
UN:1760
ADR:8,III
:GHS05
:1313-84-4