Potassium chloride 100 g/L in purified water Reag. Ph. Eur. for nitrates
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:7447-40-7
:
Formula:KCl
MDL Number:MFCD00011360
:74.55 g/mol
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless liquid
- Assay (vs. Silver Nitrate/Potassium Chromate):99-101 g/L
Specification Test Results
Frequently Bought Together