Lead stock solution 100 ppm Pb
Plastic bottle, 500 ml
Catalog # RCR4296000500A
Supplier: Ricca Chemical
Supplier #: R4296000-500A
Lead stock solution 100 ppm Pb
Catalog # RCR4296000500A
Supplier: Ricca Chemical
CAS Number: 7439-92-1
Jump to:
Product Details & Documents
- CAS Number:7439-92-1
- Formula:Pb
- MDL Number:MFCD00134050
- Molecular Weight:207.2 g/mol
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:Colorless liquid
- Lead (Pb):99.5-100.5 ppm
Specifications
- Pack type:Plastic bottle
- Size:500 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



