$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United States
ContactUsLinkComponent
Lanthanum nitrate solutions
Lanthanum nitrate solution USP Test Solution (TS)
Lanthanum nitrate solution USP Test Solution (TS)
Lanthanum nitrate solution USP Test Solution (TS)
RCR4289500500A
:
Ricca Chemical
:
Lanthanum nitrate solution USP Test Solution (TS)
RCR4289500500A
:
Ricca Chemical
:
R4289500-500A
:
Pack type:
Plastic bottle
Size:
500 ml
Cat. No.:
RCR4289500500A
Appearance:
Colorless liquid
Specific Gravity:
Actual Value Reported
:
:
Ambient