Order Entry
United States
ContactUsLinkComponent
Iodine/Potassium iodide USP Test Solution (TS) 3
Health
Iodine/Potassium iodide USP Test Solution (TS) 3
Catalog # RCR4124300500C
Supplier:  Ricca Chemical
CAS Number:  
undefined
Health
Iodine/Potassium iodide USP Test Solution (TS) 3
Catalog # RCR4124300500C
Supplier:  Ricca Chemical
Supplier Number:  R4124300-500C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    500 ml
  • Cat. No.:
    RCR4124300500C

Specifications

Specification Test Results

  • Appearance:
    Dark brown liquid
  • Assay (vs. Sodium Thiosulfate/Starch):
    12.6-12.8 g/L Iodine

Specification Test Results

About this item

 :
 :GHS08
Storage Temperature:Ambient