EDTA (ethylenediamine tetraacetic acid) 0.100 M (M/10) with Magnesium titrant
Catalog # RCR2729000-1A
Supplier: Ricca Chemical
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:1 L
- Cat. No.:RCR2729000-1A
Specifications
Specification Test Results
- Appearance:Colorless liquid
- Assay (vs. Calcium Carbonate/Eriochrome Black T):0.0999-0.1001 Molar
Specification Test Results
About this item
:
:292.25 g/mol
Storage Temperature:Ambient
CAS Number:60-00-4
Formula:C₁₀H₁₆N₂O₈
MDL Number:MFCD00003541