Order Entry
United States
ContactUsLinkComponent
Ammonium sulfide USP Test Solution (TS)
Corrosive
Oxidizing
Toxic
Ammonium sulfide USP Test Solution (TS)
Catalog # RCR0694600120C
Supplier:  Ricca Chemical
CAS Number:  
undefined
Corrosive
Oxidizing
Toxic
Ammonium sulfide USP Test Solution (TS)
Catalog # RCR0694600120C
Supplier:  Ricca Chemical
Supplier Number:  R0694600-120C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    120 ml
  • Cat. No.:
    RCR0694600120C

Specifications

Specification Test Results

  • Appearance:
    Yellow liquid
  • Suitability for Use:
    No turbidity with Magnesium Sulfate TS
  • Residue after Ignition:
    max 0.05 %

Specification Test Results

About this item

 :
 :68.14
Storage Temperature:Ambient
UN:2683
ADR:8 (6.1,3),II
 :GHS05
 :GHS03
 :GHS06
CAS Number:12135-76-1
Formula:H₈N₂S
MDL Number:MFCD00010892