DuraTube™ and Cap Assemblies, Non-Sterile
Catalog # 77506-812
Supplier: SCIENTIFIC SPECIALTIES, INC.
Specifications
- Cap included:Yes
- Capacity:2 ml
- Color:Clear
- Description:Specially formulated for bead-beating operations, non-ribbed, skirted, self-standing
- Cat. No.:77506-812
- Packaging:200/unit, 5 units/case
- Cap color:Clear
- Graduations:Graduated
Specifications
About this item
Homogenization calls for a tube that is built to withstand the rigors of lysing applications. So when you need to be sure about the tube you select for lysing and bead beating, choose the brand that has the experience, innovation, and reliability of an industry leader: SureSeal DuraTubes from SSIbio.
- Manufactured from a proprietary resin that is specially-formulated for bead-beating applications
- Shatter-resistant, they are up to the task
- Secure O-ring seal ensures a tight seal
- Ultra-pure polypropylene meets FDA 21 CFR 177.1520 & USP Class VI
- Graduated every 100 µl for easy volume estimation
- Self-standing skirted base
Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free.