Order Entry
United States
ContactUsLinkComponent
Total Ionic Strength Adjustment Buffer (TISAB II), with CDTA for Fluoride Analysis using Ion Selective Electrodes, Ricca Chemical Company
Total Ionic Strength Adjustment Buffer (TISAB II), with CDTA for Fluoride Analysis using Ion Selective Electrodes, Ricca Chemical Company
Catalog # RCR8670000-10F
Supplier:  Ricca Chemical
CAS Number:  
undefined
Total Ionic Strength Adjustment Buffer (TISAB II), with CDTA for Fluoride Analysis using Ion Selective Electrodes, Ricca Chemical Company
Catalog # RCR8670000-10F
Supplier:  Ricca Chemical
Supplier Number:  R8670000-10F
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Description:
    Total Ionic Strength Adjustment Buffer (TISAB II), with CDTA for Fluoride Analysis using Ion Selective Electrodes
  • Size:
    10 L
  • Storage Temperature:
    15°C - 30°C
  • Cat. No.:
    RCR8670000-10F

Specifications

About this item

Specification
Appearance: Colorless liquid
pH at 25°C (Method: SQCP027, SQCP033): 5.30-5.50
Suitability for use: Fluoride analysis