Buffer Concentrate, 5x, pH 7.00 ± 0.01 at 25°C (Color Coded Yellow), RICCA Chemical Company
Catalog # RCR1550510-20F
Supplier: Ricca Chemical
CAS Number:
Specifications
- Pack type:Cubitainer®
- Color:Yellow
- Description:Buffer Concentrate, 5x, pH 7.00 ± 0.01 at 25°C
- Size:20 L
- Appearance:Yellow liquid
- Reference temperature:25 °C
- Cat. No.:RCR1550510-20F
- pH range:6.990-7.010 (Method: SQCP027, SQCP033)
- Certification:ISO 17025
Specifications
About this item
5x Concentration. Ideal for limited storage space.
- Tested in ISO 17025 accredited laboratories
Certifications: ISO 17025
Packaging: 5x Concentrate: To be diluted with 4 parts water before use