ContactUsLinkComponent
Turbidity Standard, 30 NTU, Polymer Bead Suspension
  RCR8819030100C
 :  
undefined
Turbidity Standard, 30 NTU, Polymer Bead Suspension
  RCR8819030100C
 :  R8819030-100C
 :  
  • Pack type:
    Glass
  • Turbidity:
    30 NTU
  • Size:
    100 ml
  • Cat. No.:
    RCR8819030100C
Traceable to commercially prepared EPA recognized primary standards.
  • Ready to use solution
Polymer bead suspension.