Synthware Dual Vacuum Trap Assembly with #15 O-Ring Joints, Kemtech America
Catalog # 60004-610
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:Dual Trap Assembly, Complete
- Cat. No.:60004-610
- Supplier no.:V115712
Specifications
About this item
Designed for use with manifolds that have #15 O-ring joints on banks.
- Hi Vacuum valves
- Heavy wall Borosilicate construction
The pre-trap uses two 28/15 ball joints to add more flexibility to the assembly. To ensure quick connections, the pre-trap uses a 57 mm horseshoe clamp and two 57 mm O-ring flanges. A #15 O-ring joint is used to connect to the manifold. The complete assembly includes one brown VitonĀ® O-ring, two #15 O-ring clamps, and one 57 mm horseshoe clamp. Replacement components are all sold separately.