ContactUsLinkComponent
Puritan® Wound Measuring Device/DM Stick, Puritan Medical Products
Catalog # 82029-066
Product available on GSA Advantage®
undefined
Puritan® Wound Measuring Device/DM Stick, Puritan Medical Products
Catalog # 82029-066
Supplier Number:  25-15061PFDM
Product available on GSA Advantage®
Specifications
  • Sterility:
    Sterile
  • Cat. No.:
    82029-066
  • Supplier no.:
    25-1506 1PF DM
Specifications
About this item
Sticks consist of a 15.2cm (6") polystyrene shaft with an indented foam tip
Made in the USA.
The shaft has a graduated measurement scale for accurate wound measurement. Wound measurement stick 82029-066 is sterile.
Certifications: CE marked.