UNISTAT® Bilirubinometer, Reichert
Supplier: Avantor
Certificates
About this item
Bilirubinometer is a direct spectrophotometer analyzer that measures total bilirubin in neonatal serum samples
Results are provided in under five seconds, quickly determining if more extensive tests are needed. By measuring undiluted serum, errors from time-consuming dilutions or chemical reactions are eliminated. Analyzer features diagnostic codes and an expanded measuring range. It is simple to use and calibrate, and requires very little training.
Certifications: Meter is UL listed and VDE and CE marked.
Ordering information: Supplied with one-year manufacturer’s warranty, calibration cuvette, and high-level check cuvette, box of 51 disposable cuvettes, replacement bulb, reference manual, and a three-wire cord with grounded plug.
Frequently Bought Together