ContactUsLinkComponent
Synthware Photochemical Reaction Vessel with Internal Thread, Kemtech America
Synthware Photochemical Reaction Vessel with Internal Thread, Kemtech America
Catalog # 60003-762
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Photochemical Reaction Vessel with Internal Thread, Kemtech America
Catalog # 60003-762
Supplier:  Kemtech America
Supplier Number:  V150500
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • Capacity:
    500 ml
  • Cat. No.:
    60003-762
  • Supplier no.:
    V150500
Specifications
About this item
Reaction vessel is constructed of borosilicate glass to accommodate an immersion well.
  • Heavy wall Borosilicate construction
  • Ground glass joints on side arms
  • Internally threaded - #50
A #50 top internal thread accepts an RC50 bushing and Viton® O-ring to form a compression seal with the immersion well. The vessel featuers one [ST] 14/20 joint for a sparger tube, one [ST] 24/40 vertical joint for a condenser, and one #7 internal thread sidearm for a thermometer. Supplied complete with quartz immersion well, caps, and O-rings. Accessory items are sold separately.