ContactUsLinkComponent
Inoculation Loops, Disposable, Sterile
Inoculation Loops, Disposable, Sterile
Catalog # 50806-404
Supplier:  Globe Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Inoculation Loops, Disposable, Sterile
Catalog # 50806-404
Supplier:  Globe Scientific
Supplier Number:  130118
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • Capacity:
    1 µl
  • Color:
    Green
  • Cat. No.:
    50806-404
  • Supplier no.:
    130118
Specifications
About this item
Disposable loops with needles
Inoculation needle for removal of single colonies is incorporated into the reverse end of the loop for convenience and reduced costs. Designed to avoid the possibility of spreading bacteria due to the aerosol effect created by conventional flaming of reusable loops. Polystyrene loops sterilized by cobalt gamma radiation.