Order Entry
United States
ContactUsLinkComponent
 
Microbalance, XPR3
  77507-144
Microbalance, XPR3
  77507-144
 :  30875681

 

  • Model:
    XPR
  • Pan/Platform Size:
    27 mm
  • Repeatabilty:
    0.5 µg
  • Cat. No.:
    77507-144
  • Readability:
    1 µg
  • Weighing capacity:
    3.1 g

 

 

Balance XPR3 gives a supreme performance for micro samples.

  • Precise micro weighing with highest user safety
  • 3.1 g capacity
  • 1 µg readability
  • Integrated methods
  • Results notepad
  • 2 displays
  • Automatic draft shield
  • All-round visibility
  • Small footprint
  • Easy cleaning
  • LabX compatible

Exceptional accuracy for small samples:
With one microgram readability, low minimum weight, and unique design features, you can accurately weigh tiny samples with the utmost safety.

Audit-proof results - Every time:
Innovative quality assurance functions actively monitor the status of your balance and are your reassurance of accurate, repeatable, and valid results.

Small footprint and two terminals:
XPR microbalances have the smallest footprint on the market, plus the main terminal can be placed away from the balance, wherever is most convenient.

Interfaces:
Bluetooth (optional)
Ethernet (LAN)
RS232 (integrated/optional)
USB-A (to device)
USB-B (to device)