Order Entry
United States
ContactUsLinkComponent
 
Rat MD's™ Meloxicam Bacon Flavor
  76353-778
 :  Bio-Serv
Rat MD's™ Meloxicam Bacon Flavor
  76353-778
 :  Bio-Serv
 :  MD275-050

 

  • Ø:
    2.54 cm (1")
  • Height:
    0.64 cm (¼")
  • Size:
    5 g tablet
  • Cat. No.:
    76353-778
  • Packaging:
    100/bottle

 

 

Meloxicam is an effective anti-inflammatory drug used in a variety of species including rodents.

Meloxicam MD's are highly palatable and encourage appetite post-operatively promoting a more rapid recovery versus other conventional methods. They are an ideal pain-free alternative to stressful and injurious post-operative injections.

Meloxicam MD's can be customized to include antibiotics such as Baytril to provide both medications easily and affordably.

Placebo