Universal Swivel Jaw Extension Clamps
Especially adaptable for use with condensers and irregularly shaped objects
Both upper and lower swivel jaws are made of stamped steel and covered in plastisol. Lower swivel jaw, placed between the clamp arms proper and the shank, permits clamp to be oriented at any angle. Forged steel handle is 10mm (3/8") diameter. Shipping weight: 12.7kg (28lbs.).
Frequently Bought Together