ContactUsLinkComponent
Bunsen Round Jaw Extension Clamps
Bunsen Round Jaw Extension Clamps
Catalog # 21571-000
Small Business Enterprise
undefined
Bunsen Round Jaw Extension Clamps
Catalog # 21571-000
Supplier Number:  H-8562
Small Business Enterprise
Specifications
  • Length:
    203 mm (8")
  • Cat. No.:
    21571-000
  • Jaw size:
    10 - 35 mm (⅜" - 1⅜"")
  • Supplier no.:
    H-8562
Specifications
About this item
Heavy-gauge stamped steel
A concealed spring at the junction of both arms makes the jaws self-opening. With bolt and wing-nut; jaws are plastisol covered.