PYREX® Chromatographic Columns with Coarse Fritted Disc and PTFE Stopcock, PYREX®, Corning
Catalog # 89091-548
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Description:Chromatographic Extraction Apparatus with [PS] Stopcock and Fritted Disc
- Environmentally Preferable:Low Manufacturing Impact
- Environmental Certification:I-RECs, ISO 14001 - Monterrey
- Cat. No.:89091-548
- Supplier no.:38452-10
Specifications
About this item
This PYREX® column is designed for general chromatographic column separations.
- The top is tooled for a No. 00 rubber stopper fit
- A PTFE Product Standard stopcock with a No. 2 plug is provided for better column control
- Sealed into the column in a coarse porosity (40 to 60 µm) fritted disc
The volume below the disc is minimized to prevent mixing and to allow clear, sharp, band definition.