ContactUsLinkComponent
BLEACH-RITE® Plastic Test Strips, Current Technologies
BLEACH-RITE® Plastic Test Strips, Current Technologies
Catalog # 16200-186
Supplier:  Avantor
Minority or Woman-Owned Business Enterprise
undefined
BLEACH-RITE® Plastic Test Strips, Current Technologies
Catalog # 16200-186
Supplier:  Avantor
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    BLEACH-RITE® Plastic Test Strips
  • Environmentally Preferable:
  • Cat. No.:
    16200-186
  • Supplier no.:
    TS9804
Specifications
About this item
Plastic test strips change color from white to orange within 60 seconds when dipped into a bleach solution that meets the 0.525% threshold for disinfection, the 1:10 bleach-to-water dilution recommended by the CDC.