High-Performance Reflow Oven Cleaner, MicroCare
Supplier: MicroCare
Certificates
About this item
The Reflow Oven Cleaner keeps ovens at their peak performance. Made with water, alkaloids and proprietary long-chain alcohols. Cleans solder pallets, baffles, heating elements, clamps, chains, drive gears and housings, wave fingers and heat exchange units.
- Speeds cleaning of ovens even when warm
- Activated by the residual heat of the oven
- Four times faster than IPA (isopropyl alcohol)
- Cleans all solder paste and flux residues, including RMA, no-clean and lead-free products
- Non flammable for safety
- Non foaming, leaves no residue
- Low aroma
- Low GWP
Ideal for preventative maintenance. Melts away burned-on flux residues from SMT wave solder machines and reflow ovens.
Can be used in dip tanks and ultrasonic cleaners or with hand wiping. Can also be used on warm ovens (105° F).
Chemical Family: Water-based
VOC content: 50 g/L
Certifications: RoHS/WEEE/REACH/EPA SNAP compliant
Frequently Bought Together