Vesphene® IIse Non Sterile Disinfectant Cleaner, STERIS®
Catalog # 21899-870
Supplier: Steris
Specifications
- Description:Bottle
- Size:3.8 L (1 gal.)
- Cat. No.:21899-870
- Supplier no.:646108
Specifications
About this item
This one-step, alkaline, phenolic, concentrate cleaner and detergent is specifically formulated for use on hard, non porous surfaces
- Phenolic active ingredients provide broad spectrum antimicrobial efficacy.
- Formulation includes surfactants for surface cleaning while disinfecting.
- Ideal for use in a contamination control rotation.
Vesphene® IIse has a use dilution pH of 10.4 to 10.6 and is ideal for use in a disinfectant rotation program with LpH® se nonsterile disinfectant cleaner (see 16200-146).