LpH®se Nonsterile Disinfectant Cleaner, STERIS®
Supplier: Avantor
Certificates
About this item
This one-step, acid phenolic cleaner and detergent is specifically formulated for use on hard, nonporous surfaces
- Efficacy - Phenolic active ingredients provide broad spectrum antimicrobial efficacy.
- Productivity - Formulation includes surfactants for surface cleaning while disinfecting.
- Flexibility - Ideal for use in a contamination control rotation.
- Reliability - Complete lot traceability.
- Assurance - Each lot documented by Certificate of Analysis.
LpH®se has a use dilution pH of 2.6–3.0 and is ideal for use in a disinfectant rotation program with Vesphene® IIse disinfectant cleaner (see 21899-870).
Frequently Bought Together