ContactUsLinkComponent
Process NPD® st Sterile One-Step Germicidal Detergent
  89087-880
 :  Steris
Process NPD® st Sterile One-Step Germicidal Detergent
  89087-880
 :  Steris
 :  160528
  • Size:
    30 ml (1 oz.)
  • Cat. No.:
    89087-880
  • Packaging:
    Pouches, 144/case
  • Sterility:
    Sterile
  • Supplier no.:
    160528
STERIS' Process NPD® st Sterile One-Step germicidal detergent is a liquid concentrate specifically formulated for the disinfection of hard surfaces. The use-dilution can be applied manually or sprayed.
  • Efficacy - Quaternary ammonium active ingredients provide broad spectrum antimicrobial efficacy
  • Productivity - Double bagged and gamma irradiated for easy introduction into aseptic areas
  • Flexibility - Formulation includes surfactants for surface cleaning while disinfecting
  • Ease of Use - Ideal for use in a contamination control rotation
  • Assurance - Complete lot traceability and each lot documented by Certificate of Analysis
STERIS' Process NPD® st Sterile One-Step Germicidal Detergent is the sterile version of the Process NPD® family of one-step, quaternary ammonium cleaner/disinfectants made specifically for use on hard non-porous surfaces. The Process NPD® products are all made with the same formulation that does not contain perfumes or dyes that might comprise critical manufacturing systems. At a use-dilution rate of 1:256, the Process NPD® products kill a broad spectrum of common bacteria without harming the surface.