Accessories for Fyrite® Gas Analyzers, Bacharach
Supplier: Bacharach
Certificates
About this item
Repair Kits include one bottle of Fyrite fluid, valve plunger gasket, top gasket, screws, diaphragm, and envelope of filtering material.
Refill Kits include two bottles of Fyrite fluid, top gasket, screws, and envelope of filtering material.
Frequently Bought Together