Order Entry
United States
ContactUsLinkComponent
Instrument Prep Enzyme Foam, Integra™ Miltex®
Instrument Prep Enzyme Foam, Integra™ Miltex®
Catalog # 95039-236
Supplier:  Miltex
undefined
Instrument Prep Enzyme Foam, Integra™ Miltex®
Catalog # 95039-236
Supplier:  Miltex
Supplier Number:  3-760

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Bottle
  • Size:
    0.71 L (24 oz.)
  • Cat. No.:
    95039-236
  • Supplier no.:
    3-760

Specifications

About this item

Ready-to-use foaming spray for pre-cleaning of soiled instruments and scopes

Ideal for applications where instrument cleaning is not immediately available. Simply spray on instruments until ready for cleaning. Prevents bio-burden from drying onto instruments prior to decontamination. Active enzymes work to break down blood, mucus, and other bio-burden for reduced cleaning time. No water dilution is required, preventing spills and splashing during transport. Easy and safe to use on metals, plastic, and rubber materials.