Hi-Con 128 NPD Germicidal Detergent, CANI
Supplier: Cani
Certificates
About this item
Concentrated, multi-purpose, hospital-grade germicidal detergent
Effective at 8mL per liter (1oz. per gallon) in hard waters up to 400ppm, plus 5% organic serum. Detergent is pseudemonacidal, staphylocidal, salmonellacidal, fungicidal, and virucidal. Effective against HIV-1.
Frequently Bought Together