Order Entry
United States
ContactUsLinkComponent
 
7X™-O-Matic Solution, MP Biomedicals
Catalog # IC7667495
Supplier:  MP Biomedicals
undefined
7X™-O-Matic Solution, MP Biomedicals
Catalog # IC7667495
Supplier:  MP Biomedicals
Supplier Number:  097667495

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    7X-O-Matic® Cleaning Solution
  • Size:
    5 gal.
  • pH:
    9.0 - 11.0
  • Cat. No.:
    IC7667495
  • Supplier no.:
    7667495

Specifications

About this item

7X cleaning solution was developed for safe and effective cleaning of laboratory instruments and glassware.

  • Effective and safe
  • Used under pressure, heat and agitation
  • Eliminates residue leaving no interfering fluorescence
  • No etch to glass or plastic labware at any concentration
  • Completely drains from glassware in seconds
  • Works on DNase/RNase and proteases
  • Water soluable

7X-O-Matic is a powerful wetting and penetrating agent, which contains a non-ionic surfactant and a hydrotrope to complex phosphates. 7X-O-Matic was designed to clean under pressure, heat and agitation. It behaves and remains mostly suds-free when agitated. Several convenient package sizes are available, and it is stable for long-term storage.

The 7X-O-Matic® solution features alkaline salts added as builders, and is particularly suited for automatic washing machines or procedures where the solution is agitated during cleaning. It is effective at temperatures ranging from ambient to 85 °C (185 °F).

The 7X™ laboratory detergent is ideal for thorough cleaning of cultureware and general lab equipment including test tubes, petri dishes, culture flasks and cylinders, burets, pipets, slide covers, and hypodermic syringes. Solution contains powerful sequestering agents. Completely soluble in water at any concentration and drains completely from glassware in only a few seconds. A 5% solution in distilled water has a pH range of 6 to 9. This neutral pH will not induce or cause etching of delicate glassware.

Caution: Store at room temperature (15...30 °C), all 7X cleaning solutions have an indefinite shelf life in their original containers, at room temperature.