ContactUsLinkComponent
Anti-Static Wire, Justrite®
Anti-Static Wire, Justrite®
Catalog # 56610-593
Supplier:  Justrite
undefined
Anti-Static Wire, Justrite®
Catalog # 56610-593
Supplier:  Justrite
Supplier Number:  08505
Specifications
  • Description:
    Antistatic insulated wire
  • Environmentally Preferable:
  • Cat. No.:
    56610-593
  • Supplier no.:
    08505
Specifications
About this item
Prevents sparks from static electricity generated by movement and flow of flammable liquids.
Bonding between containers during liquid transfer and connecting drums to an earth ground are required under several Federal and Local Codes. Ensure adherence to these codes with this flexible 1 m (3') long insulated wire with 1.5 cm (5/8") C-clamp and 1.5 cm (5/8") alligator clip.