Glass and Laboratory Plastic Cleaner #20, Quip Laboratories
Supplier: Quip Laboratories
Certificates
About this item
Powerful liquid cleaner designed for tough cleaning problems
Effective in removing many types of laboratory markers from labware. Free rinsing leaves labware free of deposits. Sheeting action reduces spotting, filming, and drying time.
Ordering information: Cleaner can be transferred directly into your laboratory tanks through the bulk delivery service (21840-230).
Frequently Bought Together