Human Recombinant CDK2/CyclinA2 (from Baculovirus (Sf9 Insect cells))
Catalog # 77507-180
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Sf9 Insect cells)
- Species:Human
- Size:10 µg
- Tag sequence:N-terminal GST tag
- Storage Conditions:–20 °C
- Gene ID:NM_001798, NM_001237
- Protein Synonyms:CCN1|CCNA|p33(CDK2)|CDK2|CyclinA2
- Protein/Peptide Name:CDK2/CyclinA2
- Molecular Weight:~58 kDa and CyclinA2 ~78 kDa
- Formulation:50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.
- Tested Applications:Kinase assay, western blot
- Cat. No.:77507-180
Specifications
About this item
Recombinant full-length human CDK2 and CyclinA2 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins.
- Protein function: Protein kinases
Shipping temperature: Dry ice