ContactUsLinkComponent
Somso® Models of Vertebrate Hearts
Somso® Models of Vertebrate Hearts
Catalog # 470307-960
Supplier:  MARCUS SOMMER
undefined
Somso® Models of Vertebrate Hearts
Catalog # 470307-960
Supplier:  MARCUS SOMMER
Supplier Number:  ZOS54/1
Specifications
  • Description:
    Models of the hearts of vertebrates
  • Cat. No.:
    470307-960
Specifications
About this item
Somso® premium quality models.
  • Five year warranty
In SOMSO-Plast®. The internal structure is shown in all its detail. The direction of the blood flow is marked. 7 models, natural size and slightly enlarged. 1. Fish (Esox lucius), 2. Frog (Rana esculenta), 3. Tortoise (Emys orbicularis), 4. Crocodile (crocodylus niloticus), 5. Bird (Golden Eagle - Aquila chrysaetos), 6. Dog (Canis lupus familiaris) and 7. Human Being (Homo sapiens). Comprises 14 parts. Each model individually mounted on a stand with green base.
Ordering information: Items on this page are not stocked and may have an extended time for delivery.