Somso® Hand with Base of Forearm Fixed
Catalog # 470163-458
Supplier: MARCUS SOMMER
Specifications
- Description:Hand with base of forearm fixed
- Height:6 cm
- Length:30 cm
- Cat. No.:470163-458
- Width:13 cm
- Supplier no.:QS 31/2
Specifications
About this item
Somso® premium quality models.
- Five year warranty
Natural size, in SOMSO-Plast®, modelled true to nature. In one piece.
Ordering information: Items on this page are not stocked and may have an extended time for delivery.