Order Entry
United States
ContactUsLinkComponent
 
Somso® Appendix and Caecum
Catalog # 470313-130
Supplier:  MARCUS SOMMER
undefined
Somso® Appendix and Caecum
Catalog # 470313-130
Supplier:  MARCUS SOMMER
Supplier Number:  JS7

Specifications

  • Description:
    Appendix and caecum
  • Height:
    20 cm
  • Length:
    12 cm
  • Cat. No.:
    470313-130
  • Width:
    12 cm

Specifications

About this item

Somso® premium quality models.

  • Five year warranty

Natural size. In SOMSO-Plast®. The wall of the caecum can be opened. Separates into two parts. On a stand with green base.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.