ContactUsLinkComponent
Micro Sample Vial, Borosilicate Glass, with Screw Cap, Wheaton, DWK Life Sciences
Micro Sample Vial, Borosilicate Glass, with Screw Cap, Wheaton, DWK Life Sciences
Catalog # 66021-908
Small Business Enterprise
undefined
Micro Sample Vial, Borosilicate Glass, with Screw Cap, Wheaton, DWK Life Sciences
Catalog # 66021-908
Supplier Number:  225081
Small Business Enterprise
Specifications
  • Cap included:
    Yes
  • Cap Size:
    8-425
  • Cat. No.:
    66021-908
  • Supplier no.:
    225081
Specifications
About this item
A multi-purpose glass vial manufactured from Wheaton-33® low extractable borosilicate glass.
Economical when used for scintillation vial. Will contain micro samples up to 2 ml. Dimensions (with cap): 12×38 mm. With metal-foil lined white plastic cap.