ContactUsLinkComponent
Coplin Staining Jar with Cover, Wheaton
Coplin Staining Jar with Cover, Wheaton
Catalog # 25460-000
Small Business Enterprise
undefined
Coplin Staining Jar with Cover, Wheaton
Catalog # 25460-000
Supplier Number:  900470
Small Business Enterprise
Specifications
  • Dimensions:
    3 I.D. x 7.6 H cm (13/16 x 3")
  • Environmentally Preferable:
  • Cat. No.:
    25460-000
  • Supplier no.:
    900470
Specifications
About this item
Heavy soda lime glass, with ground-on cover and wide, flat base, for convenient manipulation of sections attached to slides
Holds ten 75x25mm (3x1") slides.