Order Entry
United States
ContactUsLinkComponent
Somso® Segmented Skull
Somso® Segmented Skull
Catalog # 470336-488
Supplier:  MARCUS SOMMER
Somso® Segmented Skull
Catalog # 470336-488
Supplier:  MARCUS SOMMER
Supplier Number:  QS7/8-E

Specifications

  • Description:
    Segmented skull
  • Height:
    16.5 cm
  • Length:
    22 cm
  • Cat. No.:
    470336-488
  • Width:
    15 cm

Specifications

About this item

Somso® premium quality models.

  • 5 year warranty

Developed in co-operation with Prof. Dr. med. Wolfgang Schmidt and Dr. med. Werner Scheller, institute for Anatomy, University of Leipzig. Modelled according to nature, in SOMSO-Plast®. After removing the cranium and the lower jaw, the base of the skull - sectioned medially - separates into two halves where the nasal septum, the paranasal sinuses and turbinate bones are shown. The lower jaw is movable. Separates into 5 parts. size: 51.2 cm.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.